missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540776
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PTIYRNAHDI KNKSSAPSRV PPLFVPQGTS ERKDSSGSVS PNTLSQEEGD Sequence homology: Dog: 100%; Horse: 91%; Human: 100%; Rat: 100%
PARP12 Polyclonal antibody specifically detects PARP12 in Human samples. It is validated for Western Blot.Specifications
PARP12 | |
Polyclonal | |
Unconjugated | |
PARP12 | |
9930021O16; AA409132; AA536654; ADP-ribosyltransferase diphtheria toxin-like 12; ARTD12; MST109; MSTP109; PARP12; PARP-12; poly (ADP-ribose) polymerase family, member 12; poly [ADP-ribose] polymerase 12; poly ADP-ribose polymerase 12; poly(ADP-ribose) polymerase family member 12; Protein mono-ADP-ribosyltransferase PARP12; ZC3H1; Zc3hdc1; Zinc finger CCCH domain-containing protein 1; zinc finger CCCH type domain containing 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64761 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H0J9 | |
PARP12 | |
Synthetic peptide directed towards the C-terminal of human PARP12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |