missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514158
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
PARG Polyclonal antibody specifically detects PARG in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
PARG | |
Polyclonal | |
Unconjugated | |
PARG | |
AI413217; cytosolic poly(ADP-ribose) glycohydrolase; mitochondrial poly(ADP-ribose) glycohydrolase; mitochondrial poly(ADP-ribose) glycohydrolase 63 kDa isoform; Parg; PARG99; poly (ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase 60 kDa isoform | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
8505 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q86W56 | |
PARG | |
KLH conjugated synthetic peptide between 390-421 amino acids from the C-terminal region of human Parg | |
400 μL | |
Primary | |
Human | |
Antibody | |
Ig |