missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXPAT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA146215
Additional Details : Weight : 0.01000kg
Description
The target sequence has 94% sequence homology with sheep and porcine and 84% sequence homology with rat. Suggested positive control: human and mouse heart lysate.
OXPAT Polyclonal antibody specifically detects OXPAT in Bovine, Human, Mouse, Porcine samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
OXPAT | |
Polyclonal | |
Unconjugated | |
Plin5 | |
2310076L09Rik; AI415325; AW109675; lipid droplet associated protein; lipid droplet-associated protein PAT-1; Lipid storage droplet protein 5; LSDA5; LSDP5; Mldp; Myocardial LD protein; Oxpat; Pat1; PAT-1; perilipin 5; perilipin-5; Plin5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100142670, 440503, 531199, 66968 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 30% glycerol and 0.1% sodium azide | |
A6QLL0, Q00G26, Q8BVZ1 | |
Plin5 | |
Synthetic peptide made to a region of the Human OXPAT protein, within residues 300-400. | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Porcine | |
Antibody | |
IgG |