Learn More
NRF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540912
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: WATLQGGEMT IQTTQASEAT QAVASLAEAA VAASQEMQQG ATVTMALNSE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
NRF1 Polyclonal antibody specifically detects NRF1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
NRF1 | |
Polyclonal | |
Unconjugated | |
NRF1 | |
Alpha palindromic-binding protein; alpha-pal; C87038; D6Ertd415e; EWG; LOW QUALITY PROTEIN: nuclear respiratory factor 1; Nfe2l1_retired; Not really finished protein; nrf; Nrf1; NRF-1; nrf1 protein; nrl; nuclear respiratory factor 1; nuclear respiratory factor-1; SC:BZ3C13.1; SC:bZ3C13.2; transcription factor; ube2h | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18181, 4899 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q16656, Q9WU00 | |
NRF1 | |
Synthetic peptide directed towards the middle region of human NRF1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |