missing translation for 'onlineSavingsMsg'
Learn More

NETO1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA545033

Additional Details : Weight : 0.00010kg

Product Code. 15596772

  • 472.00 € / 100µL
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Peptide sequence: MCINNTLVCN GLQNCVYPWD ENHCKEKRKT SLLDQLTNTS GTVIGVTSCI Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%

NETO1 Polyclonal antibody specifically detects NETO1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.
Specifications

Specifications

NETO1
Polyclonal
Unconjugated
NETO1
AI851453; BCTL1; brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor class A domains protein 1; Btcl1; C130005O10Rik; NETO1; neuopilin- and tolloid-like 1; neuropilin (NRP) and tolloid (TLL)-like 1; neuropilin and tolloid like 1; neuropilin- and tolloid-like protein 1; neuropilin and tolloid-like protein 1; RGD1566269; si:dkey-82k12.12
Rabbit
Affinity Chromatography
RUO
246317, 81832
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8R4I7, Q8TDF5
NETO1
Synthetic peptide directed towards the C-terminal of human NETO1
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions