missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGPRX3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533952
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla (94%) Monkey (82%).
MRGPRX3 Polyclonal antibody specifically detects MRGPRX3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
MRGPRX3 | |
Polyclonal | |
Unconjugated | |
MRGPRX3 | |
adenine receptor; G protein-coupled receptor MRGX3; G protein-coupled receptor SNSR1; G protein-coupled receptor SNSR2; GPCR; G-protein coupled receptor; G-protein coupled receptor; rc 56.1.3; Hdrr1; HDRR2; MAS related GPR family member X3; Mas-related G protein-coupled receptor g1; Mas-related gene A; mas-related GPCR member X3; MAS-related GPR, member X3; mas-related G-protein coupled receptor member A; Mas-related G-protein coupled receptor member X3; MAS-related G-protein coupled receptor, member A; MGC108580; Mgrg1; Mrga; Mrga10; Mrgpra; MRGPRX3; MRGX3; nuclear receptor MrgA10 RF-amide G protein-coupled receptor; rc 56.1.3; sensory neuron-specific G-protein coupled receptor 1/2; SNSR1; SNSR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117195 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96LB0 | |
MRGPRX3 | |
Synthetic 17 amino acid peptide from 1st extracellular domain of human MRGPRX3 / MRGX3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |