Learn More
Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin-XX, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen B2763
130.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.10000kg
Product Code. 10296492
  • 196.00 € / Each
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Description
Description
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for ChIP Assay, Immunocytochemistry, Western Blot.
Specifications
Specifications
Mouse IgG (H+L) Cross-Adsorbed
Polyclonal
Biotin-XX
Goat
purified
RUO
Mouse
Liquid
ChIP Assay, Immunocytochemistry, Western Blot
2 mg/mL
PBS with 5mM sodium azide; pH 7.5
Gamma Immunoglobins Heavy and Light chains
1 mg
Secondary
4° C