missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA125221
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A375.
MMP14 Polyclonal antibody specifically detects MMP14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
MMP14 | |
Polyclonal | |
Unconjugated | |
MMP14 | |
AI325305; cb386; EC 3.4.24.80; matrix metallo protease; matrix metallopeptidase 14; matrix metallopeptidase 14 (membrane-inserted); matrix metallopeptidase 14a (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted) alpha; matrix metalloproteinase 14 membrane-inserted; matrix metalloproteinase 14, membrane-inserted; matrix metalloproteinase 14a (membrane-inserted); matrix metalloproteinase-14; matrix metalloproteinase-14a; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; membrane-type matrix metalloproteinase 1; membrane-type matrix metalloproteinase 1 alpha; membrane-type-1 matrix metalloproteinase; MMP; MMP14; MMP-14; mmp14a; MMPs; MMP-X MT-MMP; MMP-X1; MT1MMP; MT1-MMP; MT1-MMP-alpha; Mtmmp; MT-MMP; MT-MMP 1; MT-MMP MT1-MMP; MTMMP1; MT-MMP-1; MT-MMPalpha; sabe; sb:cb386; type 1 matrix metalloprotease 14; WNCHRS; wu:fb72b09; zMT-MMPalpha; zMT-MMP-alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4323 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P50281 | |
MMP14 | |
synthetic peptide corresponding to amino acids 185-299 in the N-terminal region of human matrix metalloproteinase-14 (membrane-type matrix metalloproteinase-1), conjugated to KLH. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |