missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melan-A Antibody Cocktail, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA514017
Additional Details : Weight : 0.01000kg
Description
MA5-14017 targets MART-1 in IHC (P), IP, and WB applications and shows reactivity with Equine and Human samples. The MA5-14017 immunogen is recombinant hMART-1 protein.
Melan-A Cocktail antibody specifically detects Melan-A in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Melan-A | |
Cocktail | |
0.1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Q16655 | |
MLANA | |
Recombinant hMART-1 protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
M2-7C10, M2-9E3 | |
Unconjugated | |
MLANA | |
A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
Mouse | |
Protein A | |
RUO | |
2315 | |
4° C | |
Liquid |