missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPRE3 Monoclonal Antibody (KT36), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA172525
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with mouse EB1 or EB2.
MAPRE3 Monoclonal antibody specifically detects MAPRE3 in Human samples. It is validated for Immunocytochemistry.Specifications
MAPRE3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q9UPY8 | |
MAPRE3 | |
Recombinant human EB3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry | |
KT36 | |
Unconjugated | |
MAPRE3 | |
AI790651; APC binding protein; EB1 protein family member 3; EB2; EB3; EBF3; EBF3-S; End-binding protein 3; Mapre3; microtubule associated protein RP/EB family member 3; microtubule-associated protein RP/EB family member 3; microtubule-associated protein, RP/EB family, member 3; RP3 | |
Rat | |
Protein G | |
RUO | |
22924 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |