Learn More
Firefly luciferase Monoclonal Antibody (Luci17), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112556
Additional Details : Weight : 0.01000kg
Description
MA1-12556 detects Luciferase in C. elegans and Drosophilia melanogaster tissue, human fibroblasts, mouse macrophages, kidney, liver and cortex tissues, as well as NIH-3T3, Jurkat and BHR21 cell lines with little to no background signal.
Firefly luciferase Monoclonal antibody specifically detects Firefly luciferase in Luciferase samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
Firefly luciferase | |
Monoclonal | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
Mouse | |
Protein A/G | |
RUO | |
Luciferase | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Western Blot | |
Luci17 | |
Unconjugated | |
Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; luciferin 4-monooxygenase; Photinus pyralis; photinus pyralis luciferase | |
Luciferase protein isolated from Photinus pyralis. | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |