Learn More
ITGA2B/ITGA3, clone: 290.5C10, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121188
Additional Details : Weight : 0.01000kg
Description
The integrins are a family of more than 23 heterodimeric transmembrane proteins that mediate cell-cell adhesions as well as cell-substratum adhesions and signal transduction processes. Integrins are heterodimers consisting of noncovalently associated alpha and beta subunits. CD61 non-covalently associates with CD41 (alpha Ilb integrin) and is expressed by megakaryocytes and platelets. The CD61/CD41 complex acts as a receptor for such adhesive ligands as fibronectin, fibrinogen and von Willebrand factor during platelet stimulation.
MA1-21188 detects Integrin alpha 2b/beta 3 in Human samples.MA1-21188 has been successfully used in ELISA, FACS, Immunohistochemistry, Immunoprecipitation and Western Blot procedures.MA1-21188 immunogen corresponds to Integrin beta 3 (Human)..Specifications
Integrin alpha-2b + Integrin beta-3 | |
Monoclonal | |
Unconjugated | |
P05106, P08514 | |
ITGA2B, ITGB3 | |
Integrin beta 3 (Human). | |
RUO | |
3674, 3690 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Ascites |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
290.5C10 | |
ITGA2B | |
ITGA2B, ITGA3, CD61, GP3A, GPIIIa, Integrin alpha-Iib, GP2B, GPIIb, Platelet membrane glycoprotein Iib, CD41 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |