missing translation for 'onlineSavingsMsg'
Learn More

Influenza A M1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA532253

Additional Details : Weight : 0.01000kg


Product Code. 13426777

  • 516.00 € / 100µL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Recommended positive controls: DF-1, WSN virus infected DF-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Influenza A M1 Polyclonal antibody specifically detects Influenza A M1 in Virus samples. It is validated for Western Blot.
Specifications

Specifications

Influenza A M1
Polyclonal
Unconjugated
Influenza M1
Recombinant protein fragment of influenza A virus M1 (A/Puerto Rico/8/1934(H1N1))
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.37 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Influenza A M1 Polyclonal Antibody

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-3(2H)-isothiazolone, 2-methyl-3(2H)-isothiazolone

SDS
Documents

Documents

Certificates
Promotions

Promotions