Learn More
IL-34 Monoclonal Antibody (578416), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524259
Additional Details : Weight : 0.00010kg
Description
This antibody detects recombinant human IL-34 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-34 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <1.0 EU per 1 μg of the antibody by the LAL method.
IL-34 Monoclonal antibody specifically detects IL-34 in Human samples. It is validated for Flow Cytometry, Neutralization.Specifications
IL-34 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6ZMJ4 | |
Il34 | |
Mouse myeloma cell line NS0-derived recombinant human IL-34 Asn21-Pro242 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Neutralization | |
578416 | |
Unconjugated | |
Il34 | |
2010004A03Rik; AI593503; C16orf77; IL34; IL-34; ILN; Interleukin; interleukin 34; Interleukin34; interleukin-34 | |
Mouse | |
Protein A/G | |
RUO | |
146433 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |