Learn More
IL-2 Recombinant Polyclonal Antibody, Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 710146
Additional Details : Weight : 0.00450kg
Description
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
IL-2 Recombinant Polyclonal antibody specifically detects IL-2 in Human samples. It is validated for Western Blot.Specifications
IL-2 | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
Recombinant protein corresponding to amino acids 21–153 of human Interleukin 2 | |
100 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
2HCLC | |
Unconjugated | |
P60568 | |
Rabbit | |
Protein A | |
RUO | |
3558 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |