Learn More
IL-1 beta Monoclonal Antibody (2805), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523691
Additional Details : Weight : 0.00010kg
Description
In sandwich ELISAs, this antibody shows less than 4% cross-reactivity with recombinant rat (rr) IL-1 beta and less than 0.1% with recombinant porcine (rp) IL-1 beta, rhIL-1 alpha, rpIL-1 alpha, rrIL-1 alpha, rm IL-1 alpha, and rmIL-1 beta. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-1 beta Monoclonal antibody specifically detects IL-1 beta in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry, Neutralization, Western Blot.Specifications
IL-1 beta | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P01584, P10749 | |
IL1B | |
E. coli-derived recombinant human IL-1 beta/IL-1F2 | |
500 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Neutralization, Western Blot | |
2805 | |
Unconjugated | |
IL1B | |
catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta | |
Mouse | |
Protein A/G | |
RUO | |
16176, 3553 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |