Learn More
IFN gamma Monoclonal Antibody (CC302), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MBOIFNG
Additional Details : Weight : 0.80000kg
Product Code. 11874311
  • 554.00 € / 500µg
Estimated Shipment: 29-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
MBOIFNG targets IFN gamma in ELISA, and FACS applications and shows reactivity with Bovine, Canine, Equine, Goat, Human, Mustelid, Ovine, and Porcine samples. The MBOIFNG immunogen is recombinant Bovine IFN gamma. MBOIFNG detects IFN gamma which has a predicted molecular weight of approximately 17 kDa.
IFN gamma Monoclonal antibody specifically detects IFN gamma in Bovine, Canine, Equine, Goat, Human, Mustelid, Ovine, Porcine samples. It is validated for ELISA, Flow Cytometry.
Specifications
Specifications
IFN gamma
Monoclonal
1 mg/mL
PBS with proprietary preservative
P01579, P07353, P17773, P17803, P42160, P42161, P79154
IFNG
Recombinant Bovine IFN gamma
500 μg
Primary
Bovine, Canine, Equine, Goat, Human, Mustelid, Ovine, Porcine
Antibody
IgG1
ELISA, Flow Cytometry
CC302
Unconjugated
IFNG
BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN ?; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g
Mouse
Protein G
RUO
100034181, 100860815, 281237, 3458, 396991, 403801, 443396
-80°C