missing translation for 'onlineSavingsMsg'
Learn More

IkB alpha Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA517888

Additional Details : Weight : 0.01000kg

Product Code. 11577130

  • 424.00 € / 100µL
Estimated Shipment: 24-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It is not recommended to aliquot this antibody.

IkB alpha Polyclonal antibody specifically detects IkB alpha in Bovine, Canine, Human, Mouse, Non-human Primate, Porcine, Rat samples. It is validated for Immunoprecipitation, Western Blot.
Specifications

Specifications

IkB alpha
Polyclonal
Unconjugated
NFKBIA
AI462015; ECI-6; ECI-6/IKBA; I kappa B-alpha; I(Kappa)B(alpha); I79_018146; IkappaB alpha; IkappaBalpha; I-kappaBalpha; I-kappa-B-alpha; Ikba; IKBalpha; IKB-alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells, alpha; MAD3; MAD-3; Major histocompatibility complex enhancer-binding protein MAD3; NF kappa B inhibitor alpha; NF-kappaB inhibitor alpha; NF-kappa-B inhibitor alpha; NFKB inhibitor alpha; NFKBI; Nfkbia; nuclear factor of kappa light chain gene enhancer in B-cells; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, alpha; nuclear factor of kappa light polyp gene enhancer in B-cell 1; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha; Rel-associated pp40; REL-associated protein pp40; RL/IF-1
Rabbit
Antigen affinity chromatography
RUO
18035, 25493, 282291, 406188, 4792, 480291
-20°C
Liquid
Immunoprecipitation, Western Blot
50 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P25963, Q08353, Q63746, Q9Z1E3
NFKBIA
Synthetic peptide surrounding Arg29 of human IkB alpha.
100 μL
Primary
Bovine, Canine, Human, Mouse, Non-human Primate, Porcine, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions