missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPB8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517105
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
HSPB8 Polyclonal antibody specifically detects HSPB8 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
HSPB8 | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q9EPX0, Q9JK92, Q9UJY1 | |
HSPB8 | |
Synthetic peptide corresponding to human HSPB8/HSP22 | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
HSPB8 | |
Alpha-crystallin C chain; AU018630; AW413033; CMT2L; Cryac; crystallin, alpha C; D5Ucla4; DHMN 2; DHMN2; E2IG1; E2-induced gene 1 protein; H11; H11K; heat shock 22kDa protein 8; heat shock 27kDa protein 8; Heat shock protein; heat shock protein 8; heat shock protein B8; heat shock protein beta-8; heat shock protein family B (small) member 8; HMN 2; HMN2; HMN2A; HSB8; HSP; HSP20-like; Hsp22; HSPB 8; Hspb8; OTTHUMP00000239768; PP1629; Protein kinase H11; small stress protein-like protein HSP22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113906, 26353, 80888 | |
-20°C | |
Liquid |