missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNAI2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant GNAI2.
Brand: Abnova H00002771-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: MGCTVSAEDKAAAERSKMIDKNLREDGEKAAREVKLLLLGAGESGKSTIVKQMKIIHEDGYSEEECRQYRAVVYSNTIQSIMAIVKAMGNLQIDFADPSRADDARQLFALSCTAEEQGVLPDDLSGVIRRLWADHGVQACFGRSREYQLNDSAAYYLNDLERIAQSDYIPTQQDVLRTRVKTTGIVETHFTFKDLHFKMFDVGGQRSERKKWIHCFEGVTAIIFCVALSAYDLVLAEDEEMNRMHESMKLFDSICNNKWFTDTSIILFLNKKDLFEEKITHSPLTICFPEYTGANKYDEAASYIQSKFEDLNKRKDTKEIYTHFTCATDTKNVQFVFDAVTDVIIKNNLKDCGLFSpecifications
GNAI2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant GNAI2. | |
GNAI2 | |
GIP/GNAI2B/H_LUCA15.1/H_LUCA16.1 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC012138 | |
GNAI2 | |
GNAI2 (AAH12138, 1 a.a. ∼ 355 a.a) full-length recombinant protein with GST tag. | |
RUO | |
2771 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |