Learn More
GFR alpha-2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA118399
Additional Details : Weight : 0.01000kg
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
GFR alpha-2 Polyclonal antibody specifically detects GFR alpha-2 in Human, Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
GFR alpha-2 | |
Polyclonal | |
Unconjugated | |
GFRA2 | |
GDNF family receptor alpha 2; GDNF family receptor alpha-2; GDNF re; GDNF receptor alpha-2; GDNF receptor beta; GDNFR-alpha-2; Gdnfrb; GDNFR-beta; Gfra2; GFR-alpha 2; GFR-alpha-2; glial cell line derived neurotrophic factor family receptor alpha 2; glial cell line derived neurotrophic factor receptor, beta; HpV18gp1; neurturin receptor alpha; NRTNR-alpha; NTNRA; NTNR-alpha; PI-linked cell-surface accessory protein; RET ligand 2; RETL2; TGF-beta-related neurotrophic factor receptor 2; TRN receptor, GPI-anchored; TRNR2; tyrosine kinase receptor ligand 2 | |
Rabbit | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
O00451 | |
GFRA2 | |
Synthetic peptide corresponding to residues P(376) R V E K T P S L P D D L S D(390) of human GFR alpha-2. | |
RUO | |
25136, 2675 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |