missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADD Monoclonal Antibody (FD19), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA120168
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FADD Monoclonal antibody specifically detects FADD in Human, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Specifications
FADD | |
Monoclonal | |
2.1 mg/mL | |
0.01M PBS with 15mM sodium azide; pH 7.4 | |
Q13158 | |
FADD | |
Recombinant, full length, human FADD protein. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Western Blot | |
FD19 | |
Unconjugated | |
FADD | |
Fadd; Fas (TNFRSF6)-associated via death domain; Fas associated via death domain; FAS-associated death domain protein; FAS-associating death domain-containing protein; Fas-associating protein with death domain; GIG3; Growth-inhibiting gene 3 protein; mediator of receptor induced toxicity; mediator of receptor-induced toxicity; MGC8528; MORT1; Mort1/FADD; protein FADD | |
Mouse | |
purified | |
RUO | |
266610, 8772 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |