Learn More
Cytokeratin Pan Type I/II Antibody Cocktail, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA513156
Additional Details : Weight : 0.01000kg
Description
MA5-13156 targets Cytokeratin Pan in WB, IF and IHC (P) applications and shows reactivity with Bovine, Chicken, Human, mouse, Non-human primate, Rabbit, and Rat samples. This antibody is not suitable for PC12 cells in IF or WB applications. The MA5-13156 immunogen is human epidermal keratin. This antibody detects the acidic and basic (Type I and II) cytokeratins: Cytokeratin 1, Cytokeratin 2, Cytokeratin 3, Cytokeratin 4, Cytokeratin 5, Cytokeratin 6, Cytokeratin 7, Cytokeratin 8, Cytokeratin 10, Cytokeratin 14, Cytokeratin 15, Cytokeratin 16 and Cytokeratin 19.
Cytokeratin Pan Type I/II Cocktail antibody specifically detects Cytokeratin Pan Type I/II in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Cytokeratin Pan Type I/II | |
Cocktail | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P02533, P02535, P02538, P04104, P04264, P05783, P05784, P05787, P07744, P08727, P08729, P08730, P08779, P11679, P12035, P13645, P13646, P13647, P19001, P19012, P19013, P35908, P50446, Q01546, Q04695, Q3TTY5, Q3UV17, Q61414, Q61781, Q6IFZ6, Q7Z794, Q922U2, Q9DCV7, Q9QWL7, Q9Z2K1 | |
KRT1, KRT10, KRT13, KRT14, KRT15, KRT16, KRT17, KRT18, KRT19, KRT2, KRT3, KRT4, KRT5, Krt6a, KRT7, KRT76, KRT77, KRT8 | |
Human epidermal keratin | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
AE1/AE3 | |
Unconjugated | |
KRT19 | |
2310016L08Rik; 3300001P10Rik; 39.1; 40-kDa keratin intermediate filament; 47 kDa cytokeratin; 56 kDa cytokeratin; 57kd keratin; 57kDa keratin; 58 kDa cytokeratin; 60-kDa keratin; 63kDa Keratin; 65 kDa cytokeratin; 67 kDa cytokeratin; AA960620; adult keratin; AI324768; AI528832; AI626930; AI663979; AL022697; alpha keratin; AU019895; AW108092; AW146334; basic epidermal type II cytokeratin (carboxy-terminal region, clone pUF164); BB005427; BCIE; BIE; Card2; cell proliferation-inducing gene 46 protein; CK 2e; CK1; CK-1; CK10; CK-10; CK13; CK-13; CK14; CK-14; CK15; CK-15; CK16; CK-16; CK-17; CK18; CK-18; CK19; CK-19; CK-1B; CK-2e; CK3; CK-3; CK4; CK-4; CK5; CK-5; ck55; CK6A; CK-6A; CK6C; CK-6C; CK6D; CK-6D; CK-6E; CK7; CK-7; CK8; CK-8; CYK18; CYK4; CYK8; CYKER; cytokeratin 1; cytokeratin 10; cytokeratin 13; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 3; cytokeratin 4; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; cytokeratin 7 | |
Mouse | |
Protein G | |
RUO | |
110308, 110310, 16661, 16663, 16664, 16665, 16666, 16667, 16668, 16669, 16678, 16681, 16682, 16687, 16691, 374454, 3848, 3849, 3850, 3851, 3852, 3853, 3855, 3856, 3858, 3860, 3861, 3866, 3868, 3872, 3875, 3880, 406220, 51350, 77055 | |
4° C | |
Liquid |