Learn More
CHMP2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531128
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Mouse (100%), Xenopus laevis (93%), Pig (99%), Chicken (95%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CHMP2B Polyclonal antibody specifically detects CHMP2B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CHMP2B | |
Polyclonal | |
Unconjugated | |
Chmp2b | |
1190006E07Rik; ALS17; CGI-84; Charged multivesicular body protein 2b; charged multivesicular body protein 2Bb; CHMP family; CHMP2.5; CHMP2B; chmp2bb; chmp2bl; chromatin modifying protein 2B; chromatin modifying protein 2B, like; chromatin modifying protein 2Bb; chromatin-modifying protein 2b; DMT1; hVps2-2; hypothetical protein LOC615954; hypothetical protein zgc:77025; RGD1306781; si:ch211-106g5.3; Vacuolar protein sorting-associated protein 2-2; vacuolar protein-sorting-associated protein 2-2; VPS2 homolog B; VPS2-2; VPS2B; zgc:77025 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25978 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.59 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UQN3 | |
Chmp2b | |
Recombinant fragment corresponding to a region within amino acids 19 and 213 of Human CHMP2B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |