missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK1-A Monoclonal Antibody (3E1.1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191598
Additional Details : Weight : 0.01000kg
Description
MA1-91598 detects CDC2 from Xenopus laevis samples.
CDK1-A Monoclonal antibody specifically detects CDK1-A in Xenopus laevis samples. It is validated for Western Blot.Specifications
CDK1-A | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P35567 | |
cdk1.S | |
C terminal fragment, starting from methionine 85 of Xenopus Cdc2. | |
100 μg | |
Primary | |
X. laevis | |
Antibody | |
IgG1 |
Western Blot | |
3E1.1 | |
Unconjugated | |
cdk1.S | |
5363; ABL1; cdc; CDC2; cdc-2; cdc2 kinase; CDC28A; Cdc2a; cdc2-a; cdc2Dm; cdc2x1.1; CDCDm; CDK1; Cdk-1; cdk1.S; CDK1/CDC2; CDK1-A; cdk1-b; Cdk1-PA; CDKN1; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control; cell division control protein 2 homolog; Cell division control protein 2 homolog 1; cell division control protein 2-A; cell division cycle 2 homolog A; cell division cycle 2 like; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; cell division protein kinase 1; Cell division protein kinase 1-A; CG5363; CG5363-PA; cyclin dependent kinase; cyclin dependent kinase 1; cyclin-dependent kinase; Cyclin-dependent kinase 1; cyclin-dependent kinase 1 S homeolog; cyclin-dependent kinase 1-A; D2366; Dcdc2; dCdk1; DKFZp686L20222; Dm cdc2; Dmcdc2; DmCdk1; Dmel\CG5363; Dmel_CG5363; DNA polymerase delta catalytic subunit; DNA polymerase III; DNA-directed DNA polymerase delta POL3; group 4; HP | |
Mouse | |
IgG fraction | |
RUO | |
380246 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |