missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 Monoclonal Antibody (M38), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110290
Additional Details : Weight : 0.01000kg
Description
Western Blot: non-reducing conditions.
CD81 Monoclonal antibody specifically detects CD81 in Feline, Human, Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CD81 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P60033 | |
CD81 | |
MOLT-4 (human T-ALL cell line) | |
100 μg | |
Primary | |
Feline, Human, Rabbit | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
M38 | |
Unconjugated | |
CD81 | |
26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; Tspan28; Tspan-28 | |
Mouse | |
Protein A | |
RUO | |
101093144, 975 | |
4° C, do not freeze | |
Liquid |