missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ANG (Human) Recombinant Protein

Human ANG full-length ORF ( AAH54880, 25 a.a. - 147 a.a.) recombinant protein with GST-tag at N-terminal.

Marke:  Abnova™ H00000283-P01.10ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16181571

  • 295.00 € / 10 Mikrogramm


Verfügbar ab
15-02-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Sequence: QDNSRYTHFLTQHYDAKPQGRDDRYCESIMRRRGLTSPCKDINTFIHGNKRSIKAICENKNGNPHRENLRISKSSFQVTTCKLHGGSPWPPCQYRATAGFRNVVVACENGLPVHLDQSIFRRP
Spezifikation

Spezifikation

AAH54880
angiogenin, ribonuclease, RNase A family, 5
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANG
GST
283
Wheat germ expression system
10 μg
ALS9, HEL168, MGC22466, MGC71966, RNASE4, RNASE5
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation