missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ANG (Human) Recombinant Protein

Human ANG full-length ORF ( AAH54880, 25 a.a. - 147 a.a.) recombinant protein with GST-tag at N-terminal.

POA

Specifications

Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene ID (Entrez) 283
View More Specs

Description

Description

  • Sequence: QDNSRYTHFLTQHYDAKPQGRDDRYCESIMRRRGLTSPCKDINTFIHGNKRSIKAICENKNGNPHRENLRISKSSFQVTTCKLHGGSPWPPCQYRATAGFRNVVVACENGLPVHLDQSIFRRP
Specifications

Specifications

Store at -80°C. Aliquot to avoid repeated freezing and thawing.
283
Videos
SDS
Documents

Documents

Product Certifications