missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AMPK alpha-1/beta-1/gamma-2, His Tag Recombinant Protein

Human AMPK alpha-1/beta-1/gamma-2, His Tag Recombinant Protein

488.00 € - 7490.00 €

Specifications

Protein Length aa 10-end
Species Human
Recombinant Recombinant
Protein Tag His Tag
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15418903
View Documents Promotion Details
Invitrogen™
PV6238
10μg
488.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15428903
View Documents Promotion Details
Invitrogen™
PV6239
100μg
1678.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15438903
View Documents Promotion Details
Invitrogen™
PV6240
1mg
7490.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 09-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AMPK is activated in response to stresses that deplete cellular ATP (low glucose, hypoxia and ischemia) and via signaling pathways in response to adiponectin, leptin and CAMKKβ. Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tags.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

aa 10-end
Human
Recombinant
His Tag
Videos
SDS
Documents

Documents

Product Certifications