missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Tartrate Dibasic, purum p.a., ≥99.0% (T), Honeywell™

Catalog Number p-7120197
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
3164-29-2
C4H12N2O6
184.15
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
diazanium;(2R,3R)-2,3-dihydroxybutanedioate
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
This item is not returnable. View return policy
3164-29-2
C4H12N2O6
184.15
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
diazanium;(2R,3R)-2,3-dihydroxybutanedioate
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Purum p.a., crystallized,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystallized
Quantity 250 g
Linear Formula (NH4)2C4H4O6
Beilstein 6120352
Formula Weight 184.15g/mol
Percent Purity ≥99.0% (T)
Grade purum p.a.
Packaging Poly Bottle
Residue after Ignition ≤0.05% (as SO4)
Appearance Crystals
Density 1.601g/cu.cm
Chemical Name or Material Ammonium Tartrate Dibasic
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'