missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, MP Biomedicals™
Click to view available options
Quantity:
50 g
5 kg
Description
- Ammonium sulfate is a widely used reagent in molecular biology and chromatography.
- It is useful in the isolation and purification of enzymes and proteins.
- It is used for the precipitation or fractionation of proteins or for purification of antibodies.
- It is also useful for crystallographic analysis of nucleic acids and proteins
- Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography
- Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis.
- Also it is used for the HPLC analysis of phospholipids.
- Ammonium sulfate is an inorganic salt prepared by treating ammonia with sulfuric acid.
Specifications
Melting Point | >280°C (decomposition)(literature) |
Color | White |
pH | 5 to 6 (5% aq soln) |
Physical Form | Crystalline Powder |
Quantity | 5 kg |
Solubility Information | Soluble freely in water; insoluble in alcohol, acetone. |
Percent Purity | ≥99% |
Residue after Ignition | 8e-006 |
Density | 1.77g/mL at 25°C (Lit.) |
Grade | Ultra Puregrade |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'