Learn More
Ammonium dihydrogen phosphate, 98%
Product Code. 11408146
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
Product Code. 11408146
Brand: Thermo Scientific Alfa Aesar A15283.0C
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.
Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in acetone.
Notes
Incompatible with strong oxidizing agents, strong acids and sodium hypochlorite.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
SMILES N.OP(O)(O)=O
Specifications
Melting Point 198°C to 204°C
Quantity 10,000 g
Merck Index 14,543
Solubility Information Soluble in water. Slightly soluble in alcohol. Insoluble in acetone.
IUPAC Name phosphoric acid amine
Formula Weight 115.03
Percent Purity 98%
Odor Odorless
Density 1.803
Chemical Name or Material Ammonium dihydrogen phosphate