Learn More
Amfenac Sodium Monohydrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Amfenac Sodium Monohydrate
Specifications
Specifications
| Chemical Name or Material | Amfenac Sodium Monohydrate |
| CAS | 61618-27-7 |
| Analyte or Component Names | Amfenac Sodium Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H12 N O3 . Na . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2-Amino-3-benzoylbenzeneacetic acid sodium salt hydrate (1:1:1), Benzeneacetic acid, 2-amino-3-benzoyl-, monosodium salt, monohydrate (9CI) |
| InChI Formula | InChI=1S/C15H13NO3.Na.H2O/c16-14-11(9-13(17)18)7-4-8-12(14)15(19)10-5-2-1-3-6-10;;/h1-8H,9,16H2,(H,17,18);;1H2/q;+1;/p-1 |
| SMILES | O.[Na+].Nc1c(CC(=O)[O-])cccc1C(=O)c2ccccc2 |
| IUPAC Name | sodium;2-(2-amino-3-benzoylphenyl)acetate;hydrate |
| Show More |