Learn More
Amfenac Sodium Hydrate, MedChemExpress
Product Code. 16473387
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
Product Code. 16473387
missing translation for 'mfr': Medchem Express HY17479A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Amfenac Sodium Hydrate is a COX-2 inhibitor.
Amfenac Sodium Hydrate is a COX-2 inhibitor.
Specifications
Chemical Name or Material Amfenac Sodium Hydrate
CAS 61618-27-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H14NNaO4
Quantity 50 mg
Solubility Information DMSO : 150 mg/mL (508.01 mM; Need ultrasonic and warming)
SMILES O=C(O[Na])CC1=CC=CC(C(C2=CC=CC=C2)=O)=C1N.[H]O[H]
Molecular Weight (g/mol) 295.27
Formula Weight 295.27
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation