missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Alloferon 1
For cellular and molecular biology applications
69.82 € - 101.00 €
Specifications
Purification Method | HPLC |
---|---|
Content And Storage | -30°C to -10°C |
Format | Solid |
Molecular Weight (g/mol) | 1265.31 |
Product Type | Alloferon 1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15418419
|
Alfa Aesar™
J66879.!D |
0.5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15428419
|
Alfa Aesar™
J66879.M^ |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
HPLC | |
Solid | |
Alloferon 1 | |
His-Gly-Val-Ser-Gly-His-Gly-Gln-His-Gly-Val-His-Gly |
-30°C to -10°C | |
1265.31 | |
RUO - research use only |
For Research Use Only