missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AKT2 (PKB beta) Recombinant Human Protein

AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues.

1826.00 € - 7890.00 €

Specifications

Accession Number NP_001617
For Use With (Application) Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Formulation 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol
Molecular Weight (g/mol) 59.1kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428413
View Documents Promotion Details
Invitrogen™
PR4640B
100μg
1826.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10669434
View Documents Promotion Details
Invitrogen™
PV3975
1mg
7890.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AKT2 has been implicated in the metabolic actions of the hormone.

Specifications

Specifications

NP_001617
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
AKT Kinase Family
Recombinant
Baculovirus
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
59.1kDa
RUO - research use only
AKT2 (PKB beta)
Human
His Tag
Frozen
Videos
SDS
Documents

Documents

Product Certifications