Learn More
Testo™ Air Probe NTC
Brand:  Testo™ 0613 1712
Additional Details : Weight : 0.11000kg
Product Code. 10347273
  • 82.40 € / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Specifications
-50°C to +150°C
5 mm
115 mm