Learn More
Thermo Scientific™ Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
110.20 € - 266.00 €
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
12774388
|
Thermo Scientific™
598000000004 |
ThermoFlex™ 900 or 1400 Recirculating Chiller | Air and fluid filter kit (100 micron) |
110.20 €
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
15409549
|
Thermo Scientific™
598000000005 |
ThermoFlex™ 900 or 1400 Recirculating Chiller | Air and fluid filter kit (35 micron) |
110.20 €
1 set |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
13224244
|
Thermo Scientific™
598000000007 |
ThermoFlex™ 2500 Recirculating Chiller | Air and fluid filter kit (35 micron) |
124.80 €
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
12690795
|
Thermo Scientific™
598000000008 |
ThermoFlex™ 3500 or 5000 Recirculating Chiller | Air and fluid filter kit (100 micron) |
172.60 €
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
12620805
|
Thermo Scientific™
598000000009 |
ThermoFlex™ 3500 or 5000 Recirculating Chiller | Air and fluid filter kit (35 micron) |
213.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
12630805
|
Thermo Scientific™
598000000014 |
ThermoFlex™ 7500 or 1000 Recirculating Chiller | Air and fluid filter kit (100 micron) |
266.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
12600805
|
Thermo Scientific™
598000000013 |
ThermoFlex™ 7500 or 1000 Recirculating Chiller | Air and fluid filter kit (35 micron) |
266.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
Description
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific Neslab HX, CFT or M series recirculating chillers.