Learn More
Thermo Scientific™ Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
110.20 € - 266.00 €
Products 7
Product Code Brand For Use With (Equipment) Description Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Description Price Quantity & Availability  
12774388
View Documents Promotion Details
Thermo Scientific™
598000000004
ThermoFlex™ 900 or 1400 Recirculating Chiller Air and fluid filter kit (100 micron)
110.20 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
15409549
View Documents Promotion Details
Thermo Scientific™
598000000005
ThermoFlex™ 900 or 1400 Recirculating Chiller Air and fluid filter kit (35 micron)
110.20 €
1 set
View alternative products
Save Up to 
»
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
13224244
View Documents Promotion Details
Thermo Scientific™
598000000007
ThermoFlex™ 2500 Recirculating Chiller Air and fluid filter kit (35 micron)
124.80 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
12690795
View Documents Promotion Details
Thermo Scientific™
598000000008
ThermoFlex™ 3500 or 5000 Recirculating Chiller Air and fluid filter kit (100 micron)
172.60 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
12620805
View Documents Promotion Details
Thermo Scientific™
598000000009
ThermoFlex™ 3500 or 5000 Recirculating Chiller Air and fluid filter kit (35 micron)
213.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
12630805
View Documents Promotion Details
Thermo Scientific™
598000000014
ThermoFlex™ 7500 or 1000 Recirculating Chiller Air and fluid filter kit (100 micron)
266.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
12600805
View Documents Promotion Details
Thermo Scientific™
598000000013
ThermoFlex™ 7500 or 1000 Recirculating Chiller Air and fluid filter kit (35 micron)
266.00 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
Description
Description
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific Neslab HX, CFT or M series recirculating chillers.