missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Adipokinetic Hormone II, Locusta migratoria

315.00 € - 523.00 €

Specifications

Content And Storage Store at -20°C
Format Solid
Sensitivity Light sensitive
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15417499
SDS View Documents Promotion Details
Thermo Scientific™
J66224.MA
10 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15427499
SDS View Documents Promotion Details
Thermo Scientific™
J66224.MB
25 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Store at -20°C
Solid
Light sensitive
Videos
SDS
Documents

Documents

Product Certifications