Learn More
Buerkle™ Adapter for Ball Valve
With fine thread.
Brand: Buerkle™ 0600-0180
9.97 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.02550kg
Description
- 2 in. outer thread
- 3/4 in. inner thread
Specifications
Thread Adapter | |
Polypropylene | |
Ball Valves | |
3/4 in. Internal, 2 in. fine external |