missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ACOX1 (Human) Recombinant Protein
Human ACOX1 full-length ORF ( AAH10425, 1 a.a. - 660 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | AAH10425 |
---|---|
Gene ID (Entrez) | 51 |
Name | acyl-Coenzyme A oxidase 1, palmitoyl |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16171181
|
Abnova™
H00000051-P01.10UG |
10 μg |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16181181
|
Abnova™
H00000051-P01.25UG |
25 μg |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MNPDLRRERDSASFNPELLTHILDGSPEKTRRRREIENMILNDPDFQHEDLNFLTRSQRYEVAVRKSAIMVKKMREFGIADPDEIMWFKNFVHRGRPEPLDLHLGMFLPTLLHQATAEQQERFFMPAWNLEIIGTYAQTEMGHGTHLRGLETIATYDPETQEFILNSPTVTSIKWWPGGLGKTSNHAIVLAQLITKGKCYGLHAFIVPIREIGTHKPLPGITVGDIGPKFGYDEIDNGYLKMDNHRIPRENMLMKYAQVKPDGTYVKPLSNKLTYGTMVFVRSFLVGEAARALSKACTIAIRYSAVRHQSEMKPGEPEPQILDFQTQQYKLFPLLATAYAFQFVGAYMKETYHRINEGIGQGDLSELPELHALTAGLKAFTSWTANTGIEACRMACGGHGYSHCSGLPNIYVNFTPSCTFEGENTVMMLQTARFLMKSYDQVHSGKLVCGMVSYLNDLPSQRIQPQQVAVWPTMVDINSPESLTEAYKLRAARLVEIAAKNLQKEVIHRKSKEVAWNLTSVDLVRASEAHCHYVVVKLFSEKLLKIQDKAIQAVLRSLCLLYSLYGISQNAGDFLQGSIMTEPQITQVNQRVKELLTLIRSDAVALVDAFDFQDVTLGSVLGRYDGNVYENLFEWAKNSPLNKAEVHESYKHLKSLQSKL
Specifications
AAH10425 | |
acyl-Coenzyme A oxidase 1, palmitoyl | |
125% SDS-PAGE Stained with Coomassie Blue | |
ACOX, MGC1198, PALMCOX, SCOX | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
51 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACOX1 | |
GST |