missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, for HPLC, Gradient Grade, Reagent European Pharmacopoeia, Honeywell Burdick & Jackson™

Catalog Number p-7236055
Click to view available options
Quantity:
2.5 L
200 L
This item is not returnable. View return policy
75-05-8
C2H3N
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
ACN; Methyl cyanide
acetonitrile
CC#N
This item is not returnable. View return policy
75-05-8
C2H3N
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
ACN; Methyl cyanide
acetonitrile
CC#N
Melting Point -46°C
Color Colorless
CAS Max % 1
Density 0.7857 g/cm3 (20°C)
Boiling Point 82°C (1013 hPa)
Quantity 2.5 L
Flash Point 6°C
Linear Formula CH3CN
Beilstein 741857
Grade Far UV/Gradient
Chemical Name or Material Acetonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'