missing translation for 'onlineSavingsMsg'
Learn More

Acetone-d6, 100% (Isotopic), Thermo Scientific™

Catalog Number p-7051921
Click to view available options
Quantity:
10 Ea.
2 Ea.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
666-52-4
C3H6O
64.117
MFCD00044635
CSCPPACGZOOCGX-WFGJKAKNSA-N
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
522220
CHEBI:78217
1,1,1,3,3,3-hexadeuteriopropan-2-one
CC(=O)C
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
666-52-4
C3H6O
64.117
MFCD00044635
CSCPPACGZOOCGX-WFGJKAKNSA-N
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
522220
CHEBI:78217
1,1,1,3,3,3-hexadeuteriopropan-2-one
CC(=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 0.872 g/mL
Flash Point -17°C (1°F)
Odor Acetone-like
Refractive Index 1.3554
Quantity 2 Ea.
Melting Point -94°C
Boiling Point 56°C
UN Number UN1090
Formula Weight 64.13
Percent Purity 100%
Chemical Name or Material Acetone-d6, Isotopic
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'