missing translation for 'onlineSavingsMsg'
Learn More

ABT-239, MedChemExpress

Product Code. 16491247
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491247

Brand: Medchem Express HY12195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ABT-239 is a novel, highly efficacious, non-imidazole class of H3R antagonist and a transient receptor potential vanilloid type 1 (TRPV1) antagonist.

ABT-239 is a novel, highly efficacious, non-imidazole class of H3R antagonist and a transient receptor potential vanilloid type 1 (TRPV1) antagonist.

Specifications

Chemical Name or Material ABT-239
Physical Form Solid
CAS 460746-46-7
Color Earth Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H22N2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 100 mg/mL (302.65 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@H]1N(CCC2=CC3=C(C=CC(C4=CC=C(C#N)C=C4)=C3)O2)CCC1
Molecular Weight (g/mol) 330.42
Formula Weight 330.42
Percent Purity 98.49%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.