missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 9 mm Autosampler Vial Screw Thread Open Top Caps with Septa, 6 mm Hole

Fisherbrand™'s 9 mm screw thread caps and septa are fully compatible with all 9 mm screw thread vials and provide easy-on, easy-off convenience with just one turn.

Brand:  Fisherbrand™ 03-391-14

Encompass

 View more versions of this product

Product Code. 10002609

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Fisherbrand™'s 9 mm screw thread caps and septa are fully compatible with all 9 mm screw thread vials and provide easy-on, easy-off convenience with just one turn.

  • Fully compatible with all 9 mm Screw Thread Vials
  • Easy-on, easy-off convenience with just one turn
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Closures have the profile of a crimp or snap closure for compatibility with robotic autosamplers
  • Closures are shipped in sealed poly bags to prevent contamination during transport
Specifications

Specifications

Blue
No
100
Fisherbrand
Screw Cap
6 mm
Polypropylene
Red PTFE/White Silicone
55 Shore A
1 mm
9 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions