missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone hydrazone, 97%, Thermo Scientific™

Catalog Number p-4813293
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13629-22-6
C13H10N2
194.24
YCNUILAKOMIBAL-UHFFFAOYSA-N
9-fluorenone hydrazone, fluoren-9-one, hydrazone, fluorenone hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-one, hydrazone, fluoren-9-one hydrazone, 9h-fluoren-9-ylidenehydrazine, fluorenonhydrazon, fluoren-9-ylidenediazane, fluoren-9-ylidene-hydrazine
83627
fluoren-9-ylidenehydrazine
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN
This item is not returnable. View return policy
13629-22-6
C13H10N2
194.24
YCNUILAKOMIBAL-UHFFFAOYSA-N
9-fluorenone hydrazone, fluoren-9-one, hydrazone, fluorenone hydrazone, 9h-fluoren-9-ylidene hydrazine, 9h-fluoren-9-one, hydrazone, fluoren-9-one hydrazone, 9h-fluoren-9-ylidenehydrazine, fluorenonhydrazon, fluoren-9-ylidenediazane, fluoren-9-ylidene-hydrazine
83627
fluoren-9-ylidenehydrazine
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN
Melting Point 148.0°C to 150.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 07,I,251
Solubility Information Solubility in water: insoluble
Formula Weight 194.24
Percent Purity 97%
Chemical Name or Material 9-Fluorenone hydrazone
compliance-icons
Product Identifier
  • 9-Fluorenone hydrazone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'