missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Fluorenone-1-carbonyl chloride, 95%

Brand:  Alfa Aesar™ H36903.03

Additional Details : CAS : 55341-62-3 Weight : 0.00100kg ??product.attributeSet.Transport?? : UN number : 3261 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 15417717

  • 22.30 € / 1g


Estimated Shipment: 06-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

55341-62-3
242.66
ZNXLQKIGKKVHNF-UHFFFAOYSA-N
12209857
ClC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21
C14H7ClO2
MFCD20485912
9-oxo-9h-fluorene-1-carbonyl chloride, 3-trifluoromethylsulfanyl-benzoyl chloride, fluorene-9-one-1-carboxylic acid chloride
9-oxofluorene-1-carbonyl chloride

Specifications

9-Fluorenone-1-carbonyl chloride
Orange
1g
C14H7ClO2
UN3261
ZNXLQKIGKKVHNF-UHFFFAOYSA-N
9-oxofluorene-1-carbonyl chloride
12209857
Powder
Moisture Sensitive
126°C to 128°C
55341-62-3
MFCD20485912
9-oxo-9h-fluorene-1-carbonyl chloride, 3-trifluoromethylsulfanyl-benzoyl chloride, fluorene-9-one-1-carboxylic acid chloride
ClC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21
242.66
242.66
95%
Videos
SDS
Documents
Certificates