missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone-1-carbonyl chloride, 95%, Thermo Scientific™

Catalog Number 15427717
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
55341-62-3
C14H7ClO2
242.66
MFCD20485912
ZNXLQKIGKKVHNF-UHFFFAOYSA-N
9-oxo-9h-fluorene-1-carbonyl chloride, 3-trifluoromethylsulfanyl-benzoyl chloride, fluorene-9-one-1-carboxylic acid chloride
12209857
9-oxofluorene-1-carbonyl chloride
ClC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21
This item is not returnable. View return policy
55341-62-3
C14H7ClO2
242.66
MFCD20485912
ZNXLQKIGKKVHNF-UHFFFAOYSA-N
9-oxo-9h-fluorene-1-carbonyl chloride, 3-trifluoromethylsulfanyl-benzoyl chloride, fluorene-9-one-1-carboxylic acid chloride
12209857
9-oxofluorene-1-carbonyl chloride
ClC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 126°C to 128°C
Color Orange
Quantity 10 g
UN Number UN3261
Sensitivity Moisture Sensitive
Formula Weight 242.66
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 9-Fluorenone-1-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'