missing translation for 'onlineSavingsMsg'
Learn More

9-Aminophenanthrene, 96%, Thermo Scientific Chemicals

Catalog Number p-7052782
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
947-73-9
C14H11N
193.249
MFCD00001177
KIHQWOBUUIPWAN-UHFFFAOYSA-N
9-aminophenanthrene, 9-phenanthrenamine, 9-phenanthrylamine, phenanthren-9-ylamine, unii-7m9m5km2xy, ccris 7006, 7m9m5km2xy, 9ap, acmc-20alvr, phenanthren-9-yl-amine
13695
CHEBI:50475
phenanthren-9-amine
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)N
This item is not returnable. View return policy
947-73-9
C14H11N
193.249
MFCD00001177
KIHQWOBUUIPWAN-UHFFFAOYSA-N
9-aminophenanthrene, 9-phenanthrenamine, 9-phenanthrylamine, phenanthren-9-ylamine, unii-7m9m5km2xy, ccris 7006, 7m9m5km2xy, 9ap, acmc-20alvr, phenanthren-9-yl-amine
13695
CHEBI:50475
phenanthren-9-amine
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)N

9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.

Solubility
It is slightly soluble in water 0.00581 mg/mL.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 137°C to 139°C
Color White to Brown
Quantity 250 mg
Solubility Information It is slightly soluble in water 0.00581mg/mL.
Formula Weight 193.24
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 9-Aminophenanthrene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'