missing translation for 'onlineSavingsMsg'
Learn More

9-Acetylanthracene, 95%, Thermo Scientific Chemicals

Catalog Number p-49746
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
784-04-3
C16H12O
220.27
MFCD00001259
NXXNVJDXUHMAHU-UHFFFAOYSA-N
9-acetylanthracene, 1-anthracen-9-yl ethanone, 9-acetoanthracene, 9-anthryl methyl ketone, ethanone, 1-9-anthracenyl, 1-9-anthryl ethanone, ketone, 9-anthryl methyl, 9-acetyl anthracene, 9-acetylanthrancene, 9-acetylantracene
69911
1-anthracen-9-ylethanone
CC(=O)C1=C2C=CC=CC2=CC3=CC=CC=C31
This item is not returnable. View return policy
784-04-3
C16H12O
220.27
MFCD00001259
NXXNVJDXUHMAHU-UHFFFAOYSA-N
9-acetylanthracene, 1-anthracen-9-yl ethanone, 9-acetoanthracene, 9-anthryl methyl ketone, ethanone, 1-9-anthracenyl, 1-9-anthryl ethanone, ketone, 9-anthryl methyl, 9-acetyl anthracene, 9-acetylanthrancene, 9-acetylantracene
69911
1-anthracen-9-ylethanone
CC(=O)C1=C2C=CC=CC2=CC3=CC=CC=C31

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 72°C to 76°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 07, II, 450
Formula Weight 220.27
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 9-Acetylanthracene, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'